Learn More
Uvex™ Fleece Jacket Basic 8950-Black
These work clothes were optimised with clothing physiology in mind, and stand out thanks to their sporty lines and ergonomically placed stretch inserts. This makes for excellent wearer comfort and mobility. The design is based on our uvex protection perfect protective clothing collection - this lets you ensure that your employees dress uniformly, even for very different activities.
775.00 SEK
Specifications
Gender | Men's |
---|---|
Material | Cotton, Polyester |
Color | Black |
Closure Type | Zipper |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17206691
|
Uvex™
8940815 |
4X-Large | N/A | |||||||
17216691
|
Uvex™
8940816 |
5X-Large | N/A | |||||||
17226691
|
Uvex™
8940817 |
6X-Large | N/A | |||||||
17246681
|
Uvex™
8940809 |
Small | N/A | |||||||
17256681
|
Uvex™
8940810 |
Medium | N/A | |||||||
17266681
|
Uvex™
8940811 |
Large | N/A | |||||||
17276681
|
Uvex™
8940812 |
X-Large | N/A | |||||||
17286681
|
Uvex™
8940813 |
2X-Large |
775.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
17296681
|
Uvex™
8940814 |
3X-Large | N/A | |||||||
Description
- Material: 100% polyester
- The ideal partner all year round: Warms when worn separately at cool temperatures or in bad weather in combination with other weather jackets
- Stand-up collar and zip panel with chin guard, visible plastic zip in the front centre
- 1 breast pocket left and 2 side pockets with visible plastic zips, contrast stitching, hem width adjustable with elastic, longer back, high breathability, reflective details for better visibility
- Size: S - 6XL
- Care: washable at 30°C, do not use chlorine, do not tumble dry, iron: 1 dot, do not dry clean
Specifications
Men's | |
Black | |
Reusable |
Cotton, Polyester | |
Zipper | |
Thermal Jacket |