Learn More
Flotillin 2 Mouse anti-Human, Unconjugated, Clone: 1E11G12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66881-1-IG-150UL
 View more versions of this product
Product Code. 16879145
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The Flotillin 2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Flotillin 2. This antibody recognizes human, mouse antigen. The Flotillin 2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
Flotillin 2
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Mouse
Western Blot, Immunohistochemistry
1E11G12
Unconjugated
150 ÎĽL
2319
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Flotillin 2 Mouse anti-Human, Unconjugated, Clone: 1E11G12, Proteintech > 150 ÎĽL