Learn More
Flotillin 2 Mouse anti-Human, Unconjugated, Clone: 1E11G12, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Flotillin 2 |
---|---|
Clone | 1E11G12 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889145
|
Proteintech
66881-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879145
|
Proteintech
66881-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Flotillin 2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Flotillin 2. This antibody recognizes human, mouse antigen. The Flotillin 2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Flotillin 2 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
2319 | |
Primary |
1E11G12 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Mouse | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.