Learn More
Flotillin 2 Mouse anti-Human, Unconjugated, Clone: 1E11G12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66881-1-IG-150UL
 View more versions of this product
Product Code. 16879145
  • 4652.90 SEK / 150µL
Description
The Flotillin 2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Flotillin 2. This antibody recognizes human, mouse antigen. The Flotillin 2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
Flotillin 2
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Mouse
Western Blot, Immunohistochemistry
1E11G12
Unconjugated
150 ÎĽL
2319
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Flotillin 2 Mouse anti-Human, Unconjugated, Clone: 1E11G12, Proteintech > 150 ÎĽL