missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNTB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11507-1-AP-20UL
This item is not returnable.
View return policy
Description
ZNF24 is a new candidate transcription factor.Specifications
FNTB | |
Polyclonal | |
Unconjugated | |
FNTB | |
FNTB, FPTB, FTase beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110606, 2342, 64511 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49356, Q02293, Q8K2I1 | |
Fntb | |
FNTB Fusion Protein Ag2061 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FNTB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction