missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24285-1-AP-20UL
This item is not returnable.
View return policy
Description
Foxb1 gene ontology annotations related to this gene include anatomical structure morphogenesis; axon target recognition; cell differentiation; cell migration in diencephalon; epithelial cell differentiation involved in mammary gland alveolus development; floor plate development; hypothalamus cell migration; inferior colliculus development; lactation; mammary gland lobule development; mammillary body development; mammillothalamic axonal tract development; midbrain development; negative regulation of neuron apoptotic process; regulation of transcription by RNA polymerase II; somitogenesis; spinal cord development; telencephalon cell migration; thalamus development; urogenital system development; visual learning.Specifications
FOXB1 | |
Polyclonal | |
Unconjugated | |
FOXB1 | |
FKH5, forkhead box B1, Forkhead box protein B1, FOXB1, HFKH 5, Transcription factor FKH 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27023, 64290 | |
-20°C | |
Liquid |
Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q64732, Q99853 | |
Foxb1 | |
FOXB1 Fusion Protein Ag18079 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FOXB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction