Learn More
FOXD4L6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22081-1-AP-150UL
 View more versions of this product
Product Code. 16871625
  • 4652.90 SEK / 150µL
Description
5 FOXD4-like proteins (FOXD4L2, FOXD4L3, FOXD4L4, FOXD4L5 and FOXD4L6) are encoded by a strongly repeated region of the 9q12 chromosome region. They are very similar and it is therefore difficult to provide a clear and unambiguous protein sequence. Our sequences are in agreement with HGNC nomenclature.
Specifications
Specifications
FOXD4L6
Polyclonal
Unconjugated
FOXD4L6
forkhead box D4 like 6, Forkhead box protein D4 like 6, FOXD4 like 6, FOXD4L6
Rabbit
Antigen Affinity Chromatography
RUO
653404
-20°C
Liquid
Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3SYB3
FOXD4L6
FOXD4L6 Fusion Protein Ag17247
150 μL
Primary
Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
FOXD4L6 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated