missing translation for 'onlineSavingsMsg'
Learn More

FTSJ2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22556-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881975

  • 4721.67 SEK / 150µL
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Putative ribosomal RNA methyltransferase 2 (FTSJ2), also named as FJH1 or Protein FTSJ homolog 2 is a 246 amino acid protein, which belongs to the methyl-transferase superfamily. FTSJ2 localizes in the nucleus and is widely expressed in in muscle, placenta, and heart. FTSJ2 functions as a nucleolar RNA methyl-transferase involved in eukaryotic RNA processing and modification.
Specifications

Specifications

FTSJ2
Polyclonal
Unconjugated
MRM2
FJH1, FtsJ homolog 2 (E. coli), FTSJ2, Protein ftsJ homolog 2
Rabbit
Antigen Affinity Chromatography
RUO
29960, 68017
-20°C
Liquid
Immunoprecipitation, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9CPY0, Q9UI43
MRM2
FTSJ2 Fusion Protein Ag18410
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers