missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTSJ2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | FTSJ2 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891975
|
Proteintech
22556-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881975
|
Proteintech
22556-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Putative ribosomal RNA methyltransferase 2 (FTSJ2), also named as FJH1 or Protein FTSJ homolog 2 is a 246 amino acid protein, which belongs to the methyl-transferase superfamily. FTSJ2 localizes in the nucleus and is widely expressed in in muscle, placenta, and heart. FTSJ2 functions as a nucleolar RNA methyl-transferase involved in eukaryotic RNA processing and modification.Specifications
FTSJ2 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9CPY0, Q9UI43 | |
29960, 68017 | |
FTSJ2 Fusion Protein Ag18410 | |
Primary | |
-20°C | |
MRM2 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FJH1, FtsJ homolog 2 (E. coli), FTSJ2, Protein ftsJ homolog 2 | |
MRM2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FTSJ2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech