Learn More
Carbolite™ Furnace with Type R Temperature Sensor
Brand: Carbolite™ RHF1508-230SN+&02-301
133289.65 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Compliance to BSEN 61010 | |
8 L | |
Furnace | |
270 x 170 x 170 mm | |
170 mm | |
170 mm | |
270 mm | |
230 V | |
50/60 Hz |
301 PID Controller | |
1500°C | |
For use in quality control, heat treatment and research of ceramic materials. | |
505 mm | |
705 mm | |
675 mm | |
230 V 50/60 Hz 8000 w | |
8000 w |
Product Content Correction
Product Title
Carbolite™ Furnace with Type R Temperature Sensor > Capacity: 8L; Temperature Max: 1500C
Spot an opportunity for improvement?Share a Content Correction