missing translation for 'onlineSavingsMsg'
Learn More

GABRA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12410-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890114

  • 1355.00 SEK / 20µL
Estimated Shipment: 27-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Gamma -aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl- channel associated with the GABAA Receptor (GABAA-R) subtype. GABAA-Rs are important therapeutic targets for a range of sedative, anxiolytic, and hypnotic agents and are implicated in several diseases including epilepsy, anxiety, depression, and substance abuse. The GABAA-R is a multimeric subunit complex. To date aix alphas, four betas and four gammas, plus alternative splicing variants of some of these subunits, have been identified. Injection in oocytes or mammalian cell lines of cRNA coding for alpha- and beta-subunits results in the expression of functional GABAA-R sensitive to GABA. However, coexpression of a gamma-subunit is required for benzodiazepine modulation. The various effects of the benzodiazepines in brain may also be mediated via different alpha-subunits of the receptor. Lastly, phosphorylation of beta-subunits of the receptor has been shown to modulate GABAA-R function.
Specifications

Specifications

GABRA1
Polyclonal
Unconjugated
GABRA1
ECA4, EJM, GABA A receptor alpha 1, GABAA receptor alpha 1, GABRA1
Rabbit
Antigen Affinity Chromatography
RUO
14394, 2554, 29705
-20°C
Liquid
Immunoprecipitation, Flow Cytometry, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14867, P62812, P62813
Gabra1
GABRA1 Fusion Protein Ag3117
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers