missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12708-1-AP-20UL
This item is not returnable.
View return policy
Description
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified.Specifications
GABRA3 | |
Polyclonal | |
Unconjugated | |
GABRA3 | |
GABA A receptor alpha 3, GABAA receptor alpha 3, GABRA3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14396, 24947, 2556 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20236, P26049, P34903 | |
Gabra3 | |
GABRA3 Fusion Protein Ag3389 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GABRA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction