missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GABRA3 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890574
|
Proteintech
12708-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880574
|
Proteintech
12708-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified.Specifications
GABRA3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
P20236, P26049, P34903 | |
14396, 24947, 2556 | |
GABRA3 Fusion Protein Ag3389 | |
Primary | |
-20°C | |
GABRA3 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GABA A receptor alpha 3, GABAA receptor alpha 3, GABRA3 | |
Gabra3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GABRA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech