missing translation for 'onlineSavingsMsg'
Learn More

GABRA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12708-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16880574

  • 3665.00 SEK / 150µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified.
Specifications

Specifications

GABRA3
Polyclonal
Unconjugated
GABRA3
GABA A receptor alpha 3, GABAA receptor alpha 3, GABRA3
Rabbit
Antigen Affinity Chromatography
RUO
14396, 24947, 2556
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20236, P26049, P34903
Gabra3
GABRA3 Fusion Protein Ag3389
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers