Learn More
A&D Weighing HR Analytical Balance External Calibration
HR Analytical Balance with C-SHS patented weighing technology, which provides performance and a lean compact design.
11967.75 SEK - 15338.28 SEK
Specifications
Certifications/Compliance | GLP/GMP/GCP/ISO |
---|---|
Display Type | Reverse-backlit LCD |
Material | Stainless Steel |
Stabilization Time | 2 sec. |
Electrical Requirements | 11 VA to the AC adaptor (12 V, 1 A) |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
16611192
|
A&D Weighing
HR-250A |
252 g |
15338.28 SEK
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16661182
|
A&D Weighing
HR-100A |
102 g |
11967.75 SEK
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16681182
|
A&D Weighing
HR-150A |
152 g |
13057.86 SEK
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16631192
|
A&D Weighing
HR-251A |
62 g/252 g |
11967.75 SEK
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Robust construction, strong protection against dust & water
- Shock absorber function
- Auto power ON/OFF function
- B5 size footprint for use in narrow space
- Reverse backlit LCD display for easy to read weighing results
- SCF – Statistical calculation function
- Standard RS-232C
- Fast compact SHS sensor, response time of about 2 seconds
- Quick USB interface – no driver installation required (optional)
- LAN – Ethernet interface with WinCT-Plus software (optional)
- Easily removable large breeze break
- Built-in rechargeable NiMH battery pack (optional)
Specifications
GLP/GMP/GCP/ISO | |
Stainless Steel | |
11 VA to the AC adaptor (12 V, 1 A) |
Reverse-backlit LCD | |
2 sec. |