missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNTL5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17131-1-AP-150UL
This item is not returnable.
View return policy
Specifications
GALNTL5 | |
Polyclonal | |
Unconjugated | |
GALNTL5 | |
GalNAc T15, GALNT15, GALNTL5, pp GaNTase 15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
168391 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7Z4T8 | |
GALNTL5 | |
GALNTL5 Fusion Protein Ag10876 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GALNTL5 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction