Learn More
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Helps in maintaining a high performance chromatography system
Brand:  Trajan Scientific and Medical™ 062481
 View more versions of this product
Product Code. 15208843
  • 1750.00 SEK / 10m
Only null left
Estimated Shipment: 06-08-2025
Description
  • High quality fused silica tubing
  • Tubing protected with high temperature polyimide resin
  • Used in a wide range of GC, LC and biotechnology applications
Specifications
Specifications
Tubing
0.53 mm
10 m
GC, LC and Biotechnology
0.68 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated) > O.D. x I.D. x L: 0.68 x 0.53mm x 10m