missing translation for 'onlineSavingsMsg'
Learn More

GIPC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14822-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16883664

  • 1355.00 SEK / 20µL
Estimated Shipment: 30-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May be involved in G protein-linked signaling.
Specifications

Specifications

GIPC1
Polyclonal
Unconjugated
GIPC1
C19orf3, GIPC, GIPC1, GLUT1CBP, Hs.6454, IIP 1, NIP, RGS GAIP interacting protein, RGS19 interacting protein 1, RGS19IP1, SEMCAP, SYNECTIIN, SYNECTIN, Tax interaction protein 2, TIP 2
Rabbit
Antigen Affinity Chromatography
RUO
10755, 67903, 83823
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14908, Q9Z0G0, Q9Z254
Gipc1
GIPC1 Fusion Protein Ag6597
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers