missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIPC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12832-1-AP-20UL
This item is not returnable.
View return policy
Description
Probably interacts with SEMA5A. Expressed at highest levels in ascending colon and at moderate levels in adult kidney. Expressed at low levels in adult pancreas and at very low levels in adult liver. Expression is down-regulated in several primary tumors, such as kidney, colon and rectal tumors.Specifications
GIPC2 | |
Polyclonal | |
Unconjugated | |
GIPC2 | |
GIPC2, SEMCAP 2, SEMCAP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
365960, 54120, 54810 | |
-20°C | |
Liquid |
Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q498D9, Q8TF65, Q9Z2H7 | |
Gipc2 | |
GIPC2 Fusion Protein Ag3566 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GIPC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction