missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIPC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GIPC2 |
---|---|
Concentration | 0.61 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890754
|
Proteintech
12832-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880754
|
Proteintech
12832-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Probably interacts with SEMA5A. Expressed at highest levels in ascending colon and at moderate levels in adult kidney. Expressed at low levels in adult pancreas and at very low levels in adult liver. Expression is down-regulated in several primary tumors, such as kidney, colon and rectal tumors.Specifications
GIPC2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q498D9, Q8TF65, Q9Z2H7 | |
365960, 54120, 54810 | |
GIPC2 Fusion Protein Ag3566 | |
Primary | |
-20°C | |
GIPC2 |
0.61 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GIPC2, SEMCAP 2, SEMCAP2 | |
Gipc2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GIPC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech