missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16551-1-AP-150UL
This item is not returnable.
View return policy
Description
GLI4 is a protein coding gene.Specifications
GLI4 | |
Polyclonal | |
Unconjugated | |
GLI4 | |
GLI family zinc finger 4, GLI4, HKR4, Protein HKR4, Zinc finger protein GLI4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100043133, 2738, 500893 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10075 | |
9130023H24Rik, GLI4 | |
GLI4 Fusion Protein Ag9497 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GLI4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction