missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ GlobalMark™ Single Color Labeling Ribbons
Brand: Brady™ 076752
Additional Details : Weight : 0.31750kg
Specifications
105 mm Orange Ribbon | |
Single Color Labeling Ribbon | |
GlobalMark™ printer | |
60 m x 105 mm | |
60 m |
Orange | |
Printed text last for years outdoors and in industrial areas | |
1 Roll | |
60 m x 105 mm | |
105 mm |