missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPPB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15094-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is thought to encode a GDP-mannose pyrophosphorylase. The encoded protein catalyzes the conversion of mannose-1-phosphate and GTP to GDP-mannose, a reaction involved in the production of N-linked oligosaccharides. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
GMPPB | |
Polyclonal | |
Unconjugated | |
GMPPB | |
GMPPB, KIAA1851 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29925, 331026, 363145 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BTZ7, Q9Y5P6 | |
Gmppb | |
GMPPB Fusion Protein Ag7168 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GMPPB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction