missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPPB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GMPPB |
---|---|
Concentration | 0.49 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894104
|
Proteintech
15094-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884104
|
Proteintech
15094-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is thought to encode a GDP-mannose pyrophosphorylase. The encoded protein catalyzes the conversion of mannose-1-phosphate and GTP to GDP-mannose, a reaction involved in the production of N-linked oligosaccharides. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
GMPPB | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q8BTZ7, Q9Y5P6 | |
29925, 331026, 363145 | |
GMPPB Fusion Protein Ag7168 | |
Primary | |
-20°C | |
GMPPB |
0.49 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GMPPB, KIAA1851 | |
Gmppb | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GMPPB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech