missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10190-2-AP-20UL
This item is not returnable.
View return policy
Description
GTPase that associates with pre-60S ribosomal subunits in the nucleolus and is required for their nuclear export and maturation.Specifications
GNL2 | |
Polyclonal | |
Unconjugated | |
GNL2 | |
Autoantigen NGP 1, GNL2, HUMAUANTIG, Ngp 1, NGP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29889 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13823 | |
GNL2 | |
GNL2 Fusion Protein Ag0243 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GNL2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction