missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GNL2 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886403
|
Proteintech
10190-2-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16876403
|
Proteintech
10190-2-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GTPase that associates with pre-60S ribosomal subunits in the nucleolus and is required for their nuclear export and maturation.Specifications
GNL2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q13823 | |
29889 | |
GNL2 Fusion Protein Ag0243 | |
Primary | |
-20°C | |
GNL2 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Autoantigen NGP 1, GNL2, HUMAUANTIG, Ngp 1, NGP1 | |
GNL2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GNL2 Rabbit anti-Human, Polyclonal, Proteintech