Learn More
Goat, anti-Mouse, FITC, Polyclonal (Secondary), STEMCELL Technologies
Goat Polyclonal Antibody
Brand:  Stemcell Technologies 60138FI
Encompass_Preferred
 View more versions of this product
Product Code. 17189171
  • 4274.10 SEK / 1.50mg
This item is not returnable. View return policy
Description
Description
  • Fluorescein (FITC)-conjugated goat anti-mouse antibody reacts with the heavy chains on mouse IgG and the light chains common in most mouse immunoglobulins
  • The average molecular weight is reported to be ∼160 kDa
  • This antibody has minimal cross-reactivity to human, cow, horse, rabbit, and pig serum proteins but may react to immunoglobulins from other species
Specifications
Specifications
Immunofluorescence
FITC
1.5 mg
Mouse
Polyclonal
Goat
Secondary
Other
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Goat, anti-Mouse, FITC, Polyclonal (Secondary), STEMCELL Technologies