missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat, anti-Mouse, FITC, Polyclonal (Secondary), STEMCELL Technologies
Goat Polyclonal Antibody

POA - 4274.10 SEK
Specifications
Applications | Immunofluorescence |
---|---|
Classification | Polyclonal |
Conjugate | FITC |
Host Species | Goat |
Target Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17199171
![]() |
Stemcell Technologies
60139FI |
1 mg |
POA
1mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17189171
![]() |
Stemcell Technologies
60138FI |
1.5 mg |
4274.10 SEK
1.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Fluorescein (FITC)-conjugated goat anti-mouse antibody reacts with the heavy chains on mouse IgG and the light chains common in most mouse immunoglobulins
- The average molecular weight is reported to be ∼160 kDa
- This antibody has minimal cross-reactivity to human, cow, horse, rabbit, and pig serum proteins but may react to immunoglobulins from other species
Specifications
Immunofluorescence | |
FITC | |
Mouse |
Polyclonal | |
Goat | |
Secondary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Goat, anti-Mouse, FITC, Polyclonal (Secondary), STEMCELL Technologies