Learn More
GPT/ALT1 Mouse anti-Human, Unconjugated, Clone: 5F6B5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67531-1-IG-20UL
 View more versions of this product
Product Code. 16840216
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The GPT/ALT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GPT/ALT1. This antibody recognizes human, pig antigen. The GPT/ALT1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
GPT/ALT1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
5F6B5
Unconjugated
20 μL
2875
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GPT/ALT1 Mouse anti-Human, Unconjugated, Clone: 5F6B5, Proteintech > 20 μL