Learn More
GRIM19 Mouse anti-Human, Unconjugated, Clone: 3A10C5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67414-1-IG-20UL
 View more versions of this product
Product Code. 16880026
  • 1645.54 SEK / 20µL
Description
The GRIM19 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GRIM19. This antibody recognizes human antigen. The GRIM19 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GRIM19
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
3A10C5
Unconjugated
20 μL
51079
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GRIM19 Mouse anti-Human, Unconjugated, Clone: 3A10C5, Proteintech > 20 μL