Learn More
GRIM19 Mouse anti-Human, Unconjugated, Clone: 3A10C5, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GRIM19 |
---|---|
Clone | 3A10C5 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16880026
|
Proteintech
67414-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16870026
|
Proteintech
67414-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GRIM19 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GRIM19. This antibody recognizes human antigen. The GRIM19 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
GRIM19 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
51079 | |
Primary |
3A10C5 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.