missing translation for 'onlineSavingsMsg'
Learn More

GRIN2B Mouse anti-Human, Mouse, Rat, Clone: 1C5E12, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66565-1-IG-20UL

 View more versions of this product

Product Code. 16888555

  • 1645.54 SEK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer′s Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal domit 6-related mental retardation.
Specifications

Specifications

GRIN2B
Monoclonal
0.66 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q00960, Q01097, Q13224
GRIN2B
GRIN2B Fusion Protein Ag16718
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG1
Western Blot
1C5E12
Unconjugated
GRIN2B
GluN2B, GRIN2B, hNR3, NMDAR2B, NR2B, NR3
Mouse
Protein G
RUO
14812, 24410, 2904
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
GRIN2B Mouse anti-Human, Mouse, Rat, Clone: 1C5E12, Proteintech > 20 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.