Learn More
GRIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22398-1-AP-150UL
 View more versions of this product
Product Code. 16881915
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
GRIP1 is involved in glutamate receptors, which play an important role in neural plasticity, development and degeneration. The glutamate receptor interacting proteins, GRIP1 and GRIP2, members of the PDZ domain-containing protein family, mediate the trafficking and membrane organization of a number of transmembrane proteins. GRIP1 and GRIP2 specifically bind to the AMPA receptor subunits, GluR 2/3 and are involved in the targeting of GluR 2/3 to the synapse. GRIP1 is expressed in early development before the expression of AMPA receptors, while GRIP2 expression parallels that of AMPA receptors during later developmental stages. GRIP1 and GRIP2 may be involved in the induction of cerebellar long-term depression (LTD).
Specifications
Specifications
GRIP1
Polyclonal
Unconjugated
GRIP1
GRIP, GRIP1
Rabbit
Antigen Affinity Chromatography
RUO
23426, 74053
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.53 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q925T6, Q9Y3R0
Grip1
GRIP1 Fusion Protein Ag18071
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GRIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated