missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GRIP1 |
---|---|
Concentration | 0.53 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891915
|
Proteintech
22398-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881915
|
Proteintech
22398-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRIP1 is involved in glutamate receptors, which play an important role in neural plasticity, development and degeneration. The glutamate receptor interacting proteins, GRIP1 and GRIP2, members of the PDZ domain-containing protein family, mediate the trafficking and membrane organization of a number of transmembrane proteins. GRIP1 and GRIP2 specifically bind to the AMPA receptor subunits, GluR 2/3 and are involved in the targeting of GluR 2/3 to the synapse. GRIP1 is expressed in early development before the expression of AMPA receptors, while GRIP2 expression parallels that of AMPA receptors during later developmental stages. GRIP1 and GRIP2 may be involved in the induction of cerebellar long-term depression (LTD).Specifications
GRIP1 | |
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q925T6, Q9Y3R0 | |
23426, 74053 | |
GRIP1 Fusion Protein Ag18071 | |
Primary | |
-20°C | |
GRIP1 |
0.53 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GRIP, GRIP1 | |
Grip1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GRIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech