missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSDMC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27630-1-AP-20UL
This item is not returnable.
View return policy
Specifications
GSDMC | |
Polyclonal | |
Unconjugated | |
GSDMC | |
gasdermin C, GSDMC, MLZE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56169, 83492 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99NB5, Q9BYG8 | |
GSDMC | |
GSDMC Fusion Protein Ag26365 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GSDMC Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction