missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTZ1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14889-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the glutathione S-transferase (GSTs) super-family which encodes multifunctional enzymes important in the detoxification of electrophilic molecules, including carcinogens, mutagens, and several therapeutic drugs, by conjugation with glutathione. This enzyme also plays a significant role in the catabolism of phenylalanine and tyrosine. Thus defects in this enzyme may lead to severe metabolic disorders including alkaptonuria, phenylketonuria and tyrosinaemia. Several transcript variants of this gene encode multiple protein isoforms.Specifications
GSTZ1 | |
Polyclonal | |
Unconjugated | |
GSTZ1 | |
glutathione transferase zeta 1, GSTZ1, GSTZ1 1, MAAI, MAI, Maleylacetoacetate isomerase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14874, 2954, 681913 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43708, P57113, Q9WVL0 | |
Gstz1 | |
GSTZ1 Fusion Protein Ag6676 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GSTZ1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction