missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HADHA Mouse anti-Human, Mouse, Clone: 3E9B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60250-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes the alpha subunit of the mitochondrial trifunctional protein, which catalyzes the last three steps of mitochondrial beta-oxidation of long chain fatty acids. The mitochondrial membrane-bound heterocomplex is composed of four alpha and four beta subunits, with the alpha subunit catalyzing the 3-hydroxyacyl-CoA dehydrogenase and enoyl-CoA hydratase activities. Mutations in this gene result in trifunctional protein deficiency or LCHAD deficiency. The genes of the alpha and beta subunits of the mitochondrial trifunctional protein are located adjacent to each other in the human genome in a head-to-head orientation.Specifications
HADHA | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P40939, Q8BMS1 | |
Hadha | |
HADHA Fusion Protein Ag1211 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
3E9B1 | |
Unconjugated | |
HADHA | |
78 kDa gastrin binding protein, ECHA, HADH, HADHA, LCHAD, MTPA, TP ALPHA | |
Mouse | |
Protein G | |
RUO | |
3030, 97212 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HADHA Mouse anti-Human, Mouse, Clone: 3E9B1, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction