missing translation for 'onlineSavingsMsg'
Learn More

HADHA Mouse anti-Human, Mouse, Clone: 3E9B1, Proteintech

Mouse Monoclonal Antibody

1662.46 SEK - 4700.76 SEK

Specifications

Antigen HADHA
Clone 3E9B1
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16887285
View Documents
Proteintech
60250-1-IG-20UL
20 ÎĽL
1662.46 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16877285
View Documents
Proteintech
60250-1-IG-150UL
150 ÎĽL
4700.76 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes the alpha subunit of the mitochondrial trifunctional protein, which catalyzes the last three steps of mitochondrial beta-oxidation of long chain fatty acids. The mitochondrial membrane-bound heterocomplex is composed of four alpha and four beta subunits, with the alpha subunit catalyzing the 3-hydroxyacyl-CoA dehydrogenase and enoyl-CoA hydratase activities. Mutations in this gene result in trifunctional protein deficiency or LCHAD deficiency. The genes of the alpha and beta subunits of the mitochondrial trifunctional protein are located adjacent to each other in the human genome in a head-to-head orientation.
Specifications

Specifications

HADHA
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
78 kDa gastrin binding protein, ECHA, HADH, HADHA, LCHAD, MTPA, TP ALPHA
Hadha
IgG1
Protein G
Antibody
3E9B1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Human, Mouse
P40939, Q8BMS1
3030, 97212
HADHA Fusion Protein Ag1211
Primary
-20°C
HADHA
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HADHA Mouse anti-Human, Mouse, Clone: 3E9B1, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.