missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph™ Hei-VAP Expert Control ML G5B, EU-plug
The Hei-VAP Expert Control ML G5B adds numerous smart features to the Hei-VAP Expert.
Brand: Heidolph™ 574-02510-00
This item is not returnable.
View return policy
Description
Adds numerous smart features to the Hei-VAP Expert, as well as an overview of all parameters and the current process.- 5 in. touch display for controlling the entire system including the vacuum and the cooling temperature
- Motor lift model with transparent plastic coated G5 dry ice condenser
- For precise monitoring, the pictogram on the display also symbolizes active functions
- Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
- Setting of values either via the touch display or the separate quick access knobs
- Direct access to two favorites on the start screen
- Expandable solvent library. You can also transfer your own favorites to other devices using a USB stick
- Define and save own evaporation processes (ramps)
- Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation
- With USB and Micro SD interface for easy installation of software updates and transfer of individual favorites from one device to another

Specifications
5 L | |
Motor Lift Rotary Evaporator | |
EU-plug |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heidolph™ Hei-VAP Expert Control ML G5B, EU-plug > Type: Motor Lift Rotary Evaporator; Capacity (Metric): 5 L
Spot an opportunity for improvement?Share a Content Correction