Learn More
Heme Oxygenase-1-IN-1 hydrochloride, MedChemExpress
MedChemExpress Heme Oxygenase-1-IN-1 (Compound 2) hydrochloride is a heme oxygenase 1 (HO-1) inhibitor with an IC50 of 0.25 μM.

4014.63 SEK - 33750.96 SEK
Specifications
Chemical Name or Material | Heme Oxygenase-1-IN-1 hydrochloride |
---|---|
CAS | 1092851-70-1 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C13H16BrClN2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16465386
![]() |
Medchem Express
HY-111798A |
100 mg |
33750.96 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16450837
![]() |
Medchem Express
HY-111798A |
50 mg |
22500.64 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16447247
![]() |
Medchem Express
HY-111798A |
5 mg |
4014.63 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16414538
![]() |
Medchem Express
HY-111798A |
10 mg |
7235.68 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Heme Oxygenase-1-IN-1 (Compound 2) hydrochloride is a heme oxygenase 1 (HO-1) inhibitor with an IC50 of 0.25 μM.Specifications
Heme Oxygenase-1-IN-1 hydrochloride | |
Light Yellow | |
C13H16BrClN2 | |
BrC1=CC=C(CCCCN2C=CN=C2)C=C1.Cl | |
315.64 | |
Research | |
Cancer-programmed cell death |
1092851-70-1 | |
Research | |
DMSO : 125 mg/mL (396.02 mM; Need ultrasonic) ∣H2O : 100 mg/mL (316.82 mM; Need ultrasonic) | |
315.64 | |
99.03% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.