missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexakis(1H,1H,omegaH-perfluoroalkoxy)phosphazene, mass spectrometry standard, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
Specifications
Specifications
Density | 1.8000g/mL |
Packaging | Glass bottle |
Refractive Index | 1.3490 to 1.3570 |
Quantity | 1 g |
Specific Gravity | 1.8 |
Synonym | ultramark1621, ultramark 1621, hexa 2 h? methyl-1,3,5,2??,4??,6??-triazatriphosphinine |
Formula Weight | 2120 |
Chemical Name or Material | Hexakis(1H, 1H, ωH-perfluoroalkoxy)phosphazene |
Safety and Handling

Product Identifier
- Hexakis(1H,1H,omegaH-perfluoroalkoxy)phosphazene
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hexakis(1H,1H,omegaH-perfluoroalkoxy)phosphazene, mass spectrometry standard, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction