missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hikeshi Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20524-1-AP-20UL
This item is not returnable.
View return policy
Description
Required for organization and/or function of the secretory apparatus in Clara cells in lung.Specifications
Hikeshi | |
Polyclonal | |
Unconjugated | |
HIKESHI | |
C11orf73, Hikeshi, HSPC138, HSPC179, HSPC248, L7RN6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
293103, 51501, 67669 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q53FT3, Q5M808, Q9DD02 | |
HIKESHI | |
Hikeshi Fusion Protein Ag14376 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hikeshi Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction