missing translation for 'onlineSavingsMsg'
Learn More

Him4 Rabbit anti-C. elegans, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 51063-1-IG-20UL

 View more versions of this product

Product Code. 16866345

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Him4
Polyclonal
Unconjugated
Rabbit
Protein A
RUO
C. elegans
Antibody
IgG
ELISA
0.75 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Peptide
20 μL
Primary
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Him4 Rabbit anti-C. elegans, Polyclonal, Proteintech > 20 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.