Learn More
HISPPD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26018-1-AP-20UL
This item is not returnable.
View return policy
Description
Inositol phosphates.Specifications
HISPPD1 | |
Polyclonal | |
Unconjugated | |
PPIP5K2 | |
HISPPD1, hsVIP2, InsP6 and PP IP5 kinase 2, IP7K2, KIAA0433, PPIP5K2, VIP1 homolog 2, VIP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23262 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43314 | |
PPIP5K2 | |
HISPPD1 Fusion Protein Ag23374 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
HISPPD1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction