missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15857-1-AP-150UL
This item is not returnable.
View return policy
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transcription regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.Specifications
Histone H2B | |
Polyclonal | |
Unconjugated | |
HIST2H2BE | |
GL105, H2B, H2B.1, H2B/q, H2BFQ, HIST2H2BE, histone cluster 2, H2be, Histone H2B, Histone H2B GL105, Histone H2B type 2 E, Histone H2B.q | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
295274, 319190, 8349 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16778, Q64524 | |
Hist2h2be | |
Histone H2B Fusion Protein Ag7811 | |
150 ÎĽL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Histone H2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction