Learn More
Cytiva HiTrap Capto Lentil Lectin Product Code.: 15529244
Cytiva HiTrap Capto Lentil Lectin
Product Code. 15529244 Shop All Cytiva Products
5 x 1 mL, Pack of 5
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
Pack of 5
Product Code. 15529244
Brand: Cytiva 17548911
Capto Lentil Lectin is an affinity chromatography resin for purification of glycoproteins and other molecules containing carbohydrates such as α-D-mannose and α-D-glucose or sterically related residues.
  • Capto Lentil Lectin resin is based on a rigid high-flow agarose base matrix that enables high flow rates.
  • The product is available in bulk as well as in various high-throughput process development formats.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 x 1 mL
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Product Content Correction
Product Title
Cytiva HiTrap Capto Lentil Lectin >