Learn More
HMGB3L1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21246-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840865
  • 4721.67 SEK / 150µL
Estimated Shipment: 21-05-2024
to see stock.
Specifications
HMGB3L1
Polyclonal
Unconjugated
HMGB3P1
HMGB3P1
HMGB3L1 Fusion Protein Ag15748
150 μL
Primary
Human
Antibody
IgG
Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
HMG4L, HMGB3L1, HMGB3P1
Rabbit
Antigen Affinity Chromatography
RUO
128872
-20°C