Learn More
Hoefer™ Combs for HE 99X Max Submarine Electrophoresis Unit
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
Brand:  Hoefer™ HE91A-20-3.0
 View more versions of this product
Product Code. 12054707
  • 910.00 SEK / Each
Only null left
Estimated Shipment: 21-08-2025
Add to basket
Requires : Comb back must be ordered separately (see ordering table)
Description
Description
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
Combs can be used interchangeably with a single comb back.
Specifications
Specifications
HE99X Max Submarine Electrophoresis Unit
20
0.3 cm
0.18 in.
0.19 in.
Backless Comb
0.11 in.
70.5 ÎĽL
0.47 cm
0.5 cm
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Hoefer™ Combs for HE 99X Max Submarine Electrophoresis Unit > Comb; 20 wells; 3.0mm thick