Learn More
HPD Mouse anti-Human, Unconjugated, Clone: 3C9G2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67552-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840246
  • 1236.20 SEK / 20µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The HPD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HPD. This antibody recognizes human, mouse, rat, pig antigen. The HPD antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
HPD
Monoclonal
1000 μg/mL
Mouse
Primary
Pig, Rat, Mouse, Human
ELISA, Immunohistochemistry, Western Blot
3C9G2
Unconjugated
20 μL
3242
IgG2a