missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL2/MCP-1 Quantikine ELISA Kit

Human CCL2/MCP-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SCP00

 View more versions of this product

Product Code. 18782034

  • 44075.00 SEK / 1 set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Human CCL2/MCP-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CCL2/JE/MCP-1 in cell culture supernates (200 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul), urine (100 ul).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

31.20 - 2000pg/mL
HRP
Solid Phase Sandwich ELISA
Natural and Recombinant Human MCP-1
Human
6347
Cell Culture Supernates (200uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL),Urine (100uL)
10pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Sufficient materials to run ELISAs on at least six 96-well plates
ELISA
6 Plates
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&D Systems™ Human CCL2/MCP-1 Quantikine ELISA Kit > Six 96-well plates

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.